Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, Potassium dihydrogen phosphate/di-Sodium Hydrogen Phosphate, pH 4.0, 7.0, 10.0, Certipur™, MilliporeSigma™

Catalog No. M1990060001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
30 x 30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1990060001 30 x 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1990060001 Supplier MilliporeSigma Supplier No. 1.99006.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified reference material buffer solution for accurate pH calibration.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) pH Calibration
Quantity 30 x 30 mL
Solution Type pH Buffer
Packaging Fiber Case
Grade Analytical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.