Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution, Potassium dihydrogen phosphate/di-Sodium Hydrogen Phosphate, pH 7.0, Certipur™, MilliporeSigma™

Catalog No. M1990020001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
30 x 30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1990020001 30 x 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1990020001 Supplier MilliporeSigma Supplier No. 1.99002.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified reference material buffer solution for pH calibration, gravimetrically prepared and traceable to NIST and PTB standards.

Recommended Storage

+15°C to +25°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 7.0 (H2O, 25°C)
Boiling Point 109°C (1013hPa)
Color Colorless
Density 1.01g/3 (20°C)
For Use With (Application) pH Calibration
For Use With (Equipment) pH Meter and Electrode
pH 7
Physical Form Liquid
Quantity 30 x 30 mL
Solution Type pH Buffer
Packaging Fiber Case
Grade Certified
Chemical Name or Material Potassium Dihydrogen Phosphate/di-Sodium Hydrogen Phosphate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.