Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Solution, Potassium Hydrogen Phthalate, pH 4.0, Certipur™, MilliporeSigma™

Catalog No. M1990010001 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
30 x 30 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1990010001 30 x 30 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. M1990010001 Supplier MilliporeSigma Supplier No. 1.99001.0001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Certified reference material for the calibration of pH measuring instruments and electrodes.

Recommended Storage

+15°C to +25°C.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5

Specifications

Color Colorless
Density 1.003g/3
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.0
Physical Form Liquid
Quantity 30 x 30 mL
Solution Type pH Buffer
Packaging Fiber Case
Solubility Information Soluble
Grade Certified
Chemical Name or Material Potassium Hydrogen Phthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.