Learn More
J&H Berge Inc pH 10.010 Millesimal Buffer Solution (25 x 20 mL) Sachets
SDP
Catalog No. NC1614491
Encompass_Preferred
HI60010-02 is a premium quality pH 10.010 millesimal buffer solution that is NIST traceable and supplied with a Certificate of Analysis. Millesimal buffer solutions are designed for customers who require a resolution of 0.001 for their pH measurements. Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened. Hanna's line of millesimal buffer solutions have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened sachet. The HI60010-02 is for 25 sachets containing 20 mL of buffer.
Catalog No. NC1614491 Supplier J&H Berge Inc Supplier No. HI6001002
HI60010-02 is a premium quality pH 10.010 millesimal buffer solution that is NIST traceable and supplied with a Certificate of Analysis. Millesimal buffer solutions are designed for customers who require a resolution of 0.001 for their pH measurements. Each sachet has the lot number and expiration date stamped on it and is made of light block foil ensuring freshness each time one is opened. Hanna's line of millesimal buffer solutions have been specially formulated to have an expiration of 2 years from the date of manufacture for an unopened sachet. The HI60010-02 is for 25 sachets containing 20 mL of buffer.