Learn More
Crescent Chemical Co Inc BUFFER SUBSTANCE DULBECCO S
SDP
Supplier:  Crescent Chemical Co Inc SE47302.03
Encompass_Preferred
Buffer substance Dulbecco's 9,55 g/L Phosphate buffered saline, without calcium and magnesium; without phenol red; powder. Size - 50L Storage Conditions - +15 °C TO +30 °C Catalog Number - 47302.03
Catalog No. 50-249-124