Learn More
Cayman Chemical Buffer Supplement (10X), 15 mL
SDP
Catalog No. 502942431
Encompass_Preferred
Buffer Supplement (10X) has been tested and formulated to work exclusively with Cayman’s Senescence-Associated β-Galactosidase Staining Kit (Item No. 602010).
Catalog No. 50-294-2431 Supplier Cayman Chemical Supplier No. 40044615 ML
Buffer Supplement (10X) has been tested and formulated to work exclusively with Cayman’s Senescence-Associated β-Galactosidase Staining Kit (Item No. 602010).