Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Tablets pH 10, Lovibond™

Catalog No. 09050176 Shop All Tintometer Group Products
Change view
Click to view available options
pH:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH
09-050-176 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-050-176 Supplier Tintometer Group Supplier No. 515600BT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buffer Tablets pH 4, 7, and 10 are ideal for the fresh preparation of calibration solutions without producing large quantities of packaging material. Simply dissolve one pH buffer tablet in 20 mL of deionized water and wait 90 secs.

  • Long shelf-life
  • Blister packaging provides resistance to most climactic conditions

Specifications

For Use With (Application) Storage
For Use With (Equipment) Instruments and Electrodes
pH 10
Physical Form Tablet
Quantity 100 Pk.
Solution Type pH Buffer
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.