Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Tablets, pH 6.4, For Staining of Blood Smears, MilliporeSigma™

Catalog No. M1113730100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tablets
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1113730100 100 Tablets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1113730100 Supplier MilliporeSigma Supplier No. 1.11373.0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buffer tablets for the preparation of buffer solutions according to WEISE for staining blood smears.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Tablets, pH 6.4
Color White solid
pH 6.35 to 6.45
Physical Form Solid
Quantity 100 Tablets
Solution Type pH Buffer
Odor Odorless
Packaging Plastic can
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.