Learn More
Buffer Tablets, pH 6.8, For Staining of Blood Smears, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Buffer Tablets, pH 6.8, 100 tablets, White solid, Odorless, Plastic can, Solid, Healthcare
Specifications
Specifications
| Chemical Name or Material | Buffer Tablets, pH 6.8 |
| Color | White |
| pH | 6.8 |
| Physical Form | Solid |
| Quantity | 100 Tablets |
| Solution Type | pH Buffer |
| Odor | Odorless |
| Packaging | Plastic can |
| Grade | Not Available |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.