Learn More
Leica Biosystems Leica Biosystems Buffer Tray S, 140 mL - for VT1000 microtome (MPN 14046230132)
Catalog No. NC0289256
Encompass
Buffer Tray S is a replacement buffer tray with approximately 140 mL capacity used to collect and hold buffer solution during microtomy. The product is listed by Leica Biosystems (MPN 14046230132) and is identified by the manufacturer as compatible with the VT1000 microtome.
  • Approx. 140 mL capacity.
  • Collects and holds buffer solution during sectioning.
  • Compatible with VT1000 microtomes.
  • Intended as a replacement tray or spare part.
Catalog No. NC0289256 Supplier Leica Biosystems Supplier No. 14046230132
Buffer Tray S is a replacement buffer tray with approximately 140 mL capacity used to collect and hold buffer solution during microtomy. The product is listed by Leica Biosystems (MPN 14046230132) and is identified by the manufacturer as compatible with the VT1000 microtome.
  • Approx. 140 mL capacity.
  • Collects and holds buffer solution during sectioning.
  • Compatible with VT1000 microtomes.
  • Intended as a replacement tray or spare part.