Learn More
Ethos Biosciences Dilurex 1M Tris-HCL Buffer, 1000ml, Ethos
Catalog No. 5000574339
Encompass_Preferred
Dilurex 1M Tris-HCL 10X Buffer is a classic laboratory buffer at pH 7.4. It does not include a saline ionic component and is free to be mixed with other components as needed in your laboratory. kip the tedium of weighing powders and stocking raw materials that can expire. Our buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74339 Supplier Ethos Biosciences Supplier No. 4Z00321000
Dilurex 1M Tris-HCL 10X Buffer is a classic laboratory buffer at pH 7.4. It does not include a saline ionic component and is free to be mixed with other components as needed in your laboratory. kip the tedium of weighing powders and stocking raw materials that can expire. Our buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.