Learn More
Waters Corp Waters Tubing, Buffer, 1000 μL
Supplier: Waters Corp 700003847
This buffer tubing is designed to be placed between the syringe and the injection valve, preventing syringe contamination.
- Prevents syringe contamination.
- Connects between the syringe and injection valve.
- Supplied as one unit per package.
- Recommended for replacement as required.