Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer Variety Pack, pH 4, 7, 10, Mettler Toledo™
GSA_VA

Catalog No. 01917120 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Quantity:
6 x 500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-917-120 6 x 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-917-120 Supplier METTLER TOLEDO Supplier No. 63056116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Two 500mL bottle each of pH 4, 7, and 10 buffers

6 x 500mL, Bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer Variety Pack, pH 4, 7, 10
Melting Point 0°C
Boiling Point 100°C
Color Variety
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4, 7, 10
Physical Form Liquid
Quantity 6 x 500 mL
Solution Type Buffer
Traceability to NIST NIST Traceable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.