Learn More
Ethos Biosciences DiluRex 10X Imidazole Wash Buffer, 1000ml, Ethos
Catalog No. 5000574148
Encompass_Preferred
Dilurex 10X Imidazole wash buffer is a concentrated version of imidazole buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 7.2 working concentration by mixing 1 part 10X Imidazole wash buffer with 9 parts distilled water. Go beyond basic PBS or Tween washes with Imidazole Wash Buffer which mitigates subtle interferences from divalent cations and can reduce non-specific binding. It is particularly useful in sensitive assays where metal-ion mediated interference or subtle cross reactivities could skew results. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74148 Supplier Ethos Biosciences Supplier No. 4Z20101000
Dilurex 10X Imidazole wash buffer is a concentrated version of imidazole buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 7.2 working concentration by mixing 1 part 10X Imidazole wash buffer with 9 parts distilled water. Go beyond basic PBS or Tween washes with Imidazole Wash Buffer which mitigates subtle interferences from divalent cations and can reduce non-specific binding. It is particularly useful in sensitive assays where metal-ion mediated interference or subtle cross reactivities could skew results. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.