Learn More
Ethos Biosciences Dilurex iX Imidazole Wash+ Buffer, 1000ml, Ethos
Catalog No. 5000574348
Encompass_Preferred
Dilurex 1X Imidazole wash buffer is a ready-to-use working concentration of pH 7.2 imidazole buffered saline. Go beyond basic PBS or Tween washes with Imidazole Wash Buffer which mitigates subtle interferences from divalent cations and can reduce non-specific binding. It is particularly useful in sensitive assays where metal-ion mediated interference or subtle cross reactivities could skew results. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74348 Supplier Ethos Biosciences Supplier No. 4Z10101000
Dilurex 1X Imidazole wash buffer is a ready-to-use working concentration of pH 7.2 imidazole buffered saline. Go beyond basic PBS or Tween washes with Imidazole Wash Buffer which mitigates subtle interferences from divalent cations and can reduce non-specific binding. It is particularly useful in sensitive assays where metal-ion mediated interference or subtle cross reactivities could skew results. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.