Learn More
Ethos Biosciences Dilurex 10X TEA Wash Buffer, 100ml, Ethos
Catalog No. 5000574349
Encompass_Preferred
Dilurex 10X TEA wash buffer is a concentrated version of Triethanolamine (TEA) buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 6.8 working concentration by mixing 1 part 10X TEA wash buffer with 9 parts distilled water. Go beyond basic PBS or Tween washes with TEA Wash Buffer which, as a non-ionic surfactant, acts as a mild detergent. This is a classic lab buffer that works great in complex systems. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74349 Supplier Ethos Biosciences Supplier No. 4Z2005100
Dilurex 10X TEA wash buffer is a concentrated version of Triethanolamine (TEA) buffered saline intended as a stock solution. Save space by keeping 10x concentrate and dilute to 1x pH 6.8 working concentration by mixing 1 part 10X TEA wash buffer with 9 parts distilled water. Go beyond basic PBS or Tween washes with TEA Wash Buffer which, as a non-ionic surfactant, acts as a mild detergent. This is a classic lab buffer that works great in complex systems. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.