Learn More
Ethos Biosciences Dilurex 1X TEA Wash Buffer, 1000ml, Ethos
Catalog No. 5000574226
Encompass_Preferred
Dilurex 1X TEA wash buffer is a working concentration of Triethanolamine (TEA) buffered saline. Go beyond basic PBS or Tween washes with TEA Wash Buffer which, as a non-ionic surfactant, acts as a mild detergent. This is a classic lab buffer that works great in complex systems. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.
Catalog No. 50-005-74226 Supplier Ethos Biosciences Supplier No. 4Z20061000
Dilurex 1X TEA wash buffer is a working concentration of Triethanolamine (TEA) buffered saline. Go beyond basic PBS or Tween washes with TEA Wash Buffer which, as a non-ionic surfactant, acts as a mild detergent. This is a classic lab buffer that works great in complex systems. Skip the tedium of weighing powders and stocking raw materials that can expire. Our wash buffer is convenient, ready to use, and will save you hours in the lab, letting you focus on breakthroughs rather than buffer prep. Thousands of researchers trust Ethos Biosciences buffers for their proven track record in top-tier publications and protocols - upgrade your productivity today.