Learn More
IBA LifeSciences Buffer XT-R - Strep-TactinXT regeneration buffer; 250 ml
Catalog No. 502100133
Encompass_Preferred
Buffer XT-R is a ready-to-use Strep-TactinXT regeneration buffer containing magnesium chloride for competitive displacement of biotin from the modified biotin-binding pocket of Strep-TactinXT after the elution of Strep-tagII or Twin-Strep-tag fusion proteins. Please note that Buffer XT-R is not applicable for Strep-Tactin.
Catalog No. 50-210-0133 Supplier IBA LifeSciences Supplier No. 21045250
Buffer XT-R is a ready-to-use Strep-TactinXT regeneration buffer containing magnesium chloride for competitive displacement of biotin from the modified biotin-binding pocket of Strep-TactinXT after the elution of Strep-tagII or Twin-Strep-tag fusion proteins. Please note that Buffer XT-R is not applicable for Strep-Tactin.