Learn More
Azer Scientific Buffered Bluing Reagent, iCON™ Staining System
SDP
Catalog No. 21377143
Change view
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-143 1 gal.
21-377-145 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-377-143 Supplier Azer Scientific Supplier No. ES365711G
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Standard staining protocol, only thirty seconds to one minute exposure time
  • Buffered to a pH of 8
  • Provides consistent and rapid bluing of hematoxylin-stained nuclei
  • Enhances nuclear chromatin detail and initiates an insoluble dye-lake complex
  • Universal and may be used with progressive and regressive hematoxylin stains
  • Clear colorless solution
  • Two year shelf life
Specifications
pH 8
Quantity 1 gal.