Learn More
Alkali Scientific Buffered Charcoal Yeast Extract Agar (BCYE), 15x100mm, 25mL, 10/pk
SDP
Catalog No. 5000662640
Encompass_Preferred
Buffered Charcoal Yeast Extract (BCYE) Agar, 15×100mm plates containing 25 mL of medium, is an enriched selective culture medium designed for the isolation and cultivation of Legionella species and other fastidious microorganisms. The formulation includes yeast extract to provide essential nutrients and activated charcoal to neutralize toxic compounds that may inhibit bacterial growth. The buffered system helps maintain an optimal pH environment that supports consistent microbial recovery. BCYE agar is widely used in clinical diagnostics, environmental microbiology, and water testing laboratories for the detection of Legionella in environmental and clinical samples. Each plate is prepared with sterile agar poured to a uniform depth, ensuring reliable performance, consistent microbial growth, and dependable results for routine laboratory testing and research applications.
Catalog No. 50-006-62640 Supplier Alkali Scientific Supplier No. BCYEP00
Buffered Charcoal Yeast Extract (BCYE) Agar, 15×100mm plates containing 25 mL of medium, is an enriched selective culture medium designed for the isolation and cultivation of Legionella species and other fastidious microorganisms. The formulation includes yeast extract to provide essential nutrients and activated charcoal to neutralize toxic compounds that may inhibit bacterial growth. The buffered system helps maintain an optimal pH environment that supports consistent microbial recovery. BCYE agar is widely used in clinical diagnostics, environmental microbiology, and water testing laboratories for the detection of Legionella in environmental and clinical samples. Each plate is prepared with sterile agar poured to a uniform depth, ensuring reliable performance, consistent microbial growth, and dependable results for routine laboratory testing and research applications.