Learn More
Rainbow Scientific BUFFERED HYPOTONIC SOL.
SDP
Catalog No. 501092830
Encompass
It has been found to achieve good metaphase swelling (spreading) from a wide range of sample types and significantly increases the number of useful metaphases in leukaemic bone marrow cultures when compared to the standard 0.075M KCL hypotonic. It contains 0.4% KCL with HEPES.
Catalog No. 50-109-2830 Supplier Rainbow Scientific Supplier No. GGSJL006/A
It has been found to achieve good metaphase swelling (spreading) from a wide range of sample types and significantly increases the number of useful metaphases in leukaemic bone marrow cultures when compared to the standard 0.075M KCL hypotonic. It contains 0.4% KCL with HEPES.