Learn More
Alkali Scientific Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 25mL, 50/Case
SDP
Catalog No. 5000663196
Encompass_Preferred
Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 25mL, 50/Case. This high-quality pre-enrichment media is perfect for microbiological testing, offering a sterile and reliable solution. The BPW 2X formula is designed to support microbial growth by enriching the sample, making it ideal for food and water testing in laboratory environments. It comes ready to use, ensuring quick and accurate sample preparation.
Catalog No. 50-006-63196 Supplier Alkali Scientific Supplier No. BPW2X025
Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 25mL, 50/Case. This high-quality pre-enrichment media is perfect for microbiological testing, offering a sterile and reliable solution. The BPW 2X formula is designed to support microbial growth by enriching the sample, making it ideal for food and water testing in laboratory environments. It comes ready to use, ensuring quick and accurate sample preparation.