Learn More
Alkali Scientific Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 30mL, 50/Case
SDP
Catalog No. 5000662621
Encompass_Preferred
Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 30mL, 50/Case. This pre-enrichment media is ideal for use in microbiological applications, offering a sterile and ready-to-use solution. BPW 2X supports the growth of microorganisms, ensuring high accuracy in laboratory testing. Perfect for food safety and environmental microbiological testing, it helps enrich samples for further analysis.
Catalog No. 50-006-62621 Supplier Alkali Scientific Supplier No. BPW2X050
Buffered Peptone Water 2X (BPW 2X), Sterile, Pre Enrichment Media, 30mL, 50/Case. This pre-enrichment media is ideal for use in microbiological applications, offering a sterile and ready-to-use solution. BPW 2X supports the growth of microorganisms, ensuring high accuracy in laboratory testing. Perfect for food safety and environmental microbiological testing, it helps enrich samples for further analysis.