Learn More
Cole-Parmer Cole-Parmer Buffered Peptone Water, 500 g
Supplier:  Cole-Parmer 1420208
Encompass_Preferred
Helps in the recovery of Salmonella from foods after preservation techniques. Offered in the most popular formulations, our media is thoroughly tested and subject to strict examination and standardization to ensure reliable and consistent lot-to-lot performance. Contact one of our Application Specialists to find out the availability of other dehydrated culture media and learn about our custom blending service. Each order includes a certificate of compliance and quality control that shows lot numbers, expiration dates, and a shelf-life guarantee. Wide mouth openings on bottles let you scoop out powder efficiently without dusting the countertop. The lightweight plastic bottles have indented hand grips for steady handling. Wide, flat cap allows you to stack containers and save space. Media are formulated to dissolve quickly.
Catalog No. NC0525831