Learn More
Alkali Scientific Buffered Peptone Water (BPW), Sterile, Ready to Use Pre Enrichment Media, 800mL Bottle, 1/Case
SDP
Catalog No. 5000663072
Encompass_Preferred
Buffered Peptone Water (BPW), Sterile, Ready to Use Pre Enrichment Media, 800mL Bottle, 1/Case. This 800mL bottle of buffered peptone water is a key component for enriching microbiological samples before further testing. The sterile environment ensures that your samples remain uncontaminated, providing reliable and accurate results. It is widely used in laboratory environments for microbial research and testing, making it an essential product for microbiologists and scientists.
Catalog No. 50-006-63072 Supplier Alkali Scientific Supplier No. BPW800
Buffered Peptone Water (BPW), Sterile, Ready to Use Pre Enrichment Media, 800mL Bottle, 1/Case. This 800mL bottle of buffered peptone water is a key component for enriching microbiological samples before further testing. The sterile environment ensures that your samples remain uncontaminated, providing reliable and accurate results. It is widely used in laboratory environments for microbial research and testing, making it an essential product for microbiologists and scientists.