Learn More
HIMEDIA Buffered Peptone Water, M614, Microbiology Culture Media, Dehydrated Culture Media, Pre-Enrichment Broth, Salmonella Recovery, Food Testing, Powder, 500 g
Catalog No. NC4013766
Encompass_Preferred
Buffered Peptone Water is the standard non-selective pre-enrichment broth for recovering injured Salmonella from foods before selective enrichment and isolation. Proteose peptone supplies gentle nutrition while a disodium and potassium phosphate buffer system holds a stable pH near 7.2, protecting sub-lethally damaged cells as they resuscitate and multiply. This first-step broth underpins reliable Salmonella detection in food and environmental testing. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013766 Supplier HIMEDIA Supplier No. M614500G
Buffered Peptone Water is the standard non-selective pre-enrichment broth for recovering injured Salmonella from foods before selective enrichment and isolation. Proteose peptone supplies gentle nutrition while a disodium and potassium phosphate buffer system holds a stable pH near 7.2, protecting sub-lethally damaged cells as they resuscitate and multiply. This first-step broth underpins reliable Salmonella detection in food and environmental testing. Dehydrated powder, store in a cool dry place. Packaged as 500 g.