Learn More
MilliporeSigma Buff. NaCl-Pept. Sol. acc EP
Catalog No. NC1480111
Encompass
Application: Microbial examination of non-sterile pharmaceutical products, Dilution, Rinse Fluid. Packaging: 100 ml in 125 ml bottle with blue screw cap and 3 loci (10 bottles per box). Synonym: Buffered Saline Peptone Water, Buffered Sodium Chloride Peptone Solution.
Catalog No. NC1480111 Supplier MilliporeSigma Supplier No. 1463670010
Application: Microbial examination of non-sterile pharmaceutical products, Dilution, Rinse Fluid. Packaging: 100 ml in 125 ml bottle with blue screw cap and 3 loci (10 bottles per box). Synonym: Buffered Saline Peptone Water, Buffered Sodium Chloride Peptone Solution.