Learn More
Teknova Buffered Sodium Chloride-Peptone (0.19%) Solution, pH 7.0. 1000mL, Sterile.
SDP
Catalog No. 502388084
Encompass
0.19% Peptone,0.43% Sodium Chloride,0.36% Potassium Phosphate,0.57% Sodium Phosphate
Catalog No. 50-238-8084 Supplier Teknova Supplier No. S0266
0.19% Peptone,0.43% Sodium Chloride,0.36% Potassium Phosphate,0.57% Sodium Phosphate