Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Buffered Zinc Formalin
GSA_VASDP

Catalog No. STLBFZ1
Change view
Click to view available options
Packaging Type:
Bottle
Cube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Type
STLBFZ1 Bottle
STLBFZ5 Cube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
Catalog No. STLBFZ1 Supplier StatLab™ Supplier No. BFZ1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

Ready-to-use buffered zinc formalin in bulk packaging.

  • Especially recommended for use in immunohistochemistry
  • Preserves more immunoreactivity than standard NBF

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Packaging Type Bottle
Quantity 4 x 1 gal.
Formulation Formaldehyde, Zinc Sulfate Heptahydrate, Methyl Alcohol
For Use With (Application) Immunohistochemistry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.