Learn More
Cytek Biosciences Inc Buffers and Solutions Fixation Buffer ….. Cat. No. TNB-8222-L100 ….. Size 100 mL
Catalog No. 502103678
Encompass_Preferred
Buffers and Solutions Fixation Buffer ….. Cat. No. TNB-8222-L100 ….. Size 100 mLFixation Buffer can be used to fix cells prior to permeabilization when performing intracellular staining for antigens expressed on cytoplasmic proteins. This buffer can also be used to fix cells after immunofluorescent staining when the acquisition of samples will be delayed.
Catalog No. 50-210-3678 Supplier Cytek Biosciences Inc Supplier No. TNB8222L100
Buffers and Solutions Fixation Buffer ….. Cat. No. TNB-8222-L100 ….. Size 100 mLFixation Buffer can be used to fix cells prior to permeabilization when performing intracellular staining for antigens expressed on cytoplasmic proteins. This buffer can also be used to fix cells after immunofluorescent staining when the acquisition of samples will be delayed.