Learn More
BRANSON ULTRASONICS CORPORATION (3P) BUFFING COMPOUND REMOVER 12/CS
Catalog No. NC3721575
Encompass_Preferred
Branson Buffing Compound Cleaning Solution, mixed in a concentration of 6 to 7% by volume, formulated to remove buffing and polishing compounds.
Catalog No. NC3721575 Supplier BRANSON ULTRASONICS CORPORATION (3P) Supplier No. 000955314
Branson Buffing Compound Cleaning Solution, mixed in a concentration of 6 to 7% by volume, formulated to remove buffing and polishing compounds.