Learn More
Apexbio Technology LLC Buflomedil HCl 35543-24-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000092231
Encompass_Preferred
Buflomedil HCl is a peripheral vasodilating agent targeting alpha-adrenergic receptors It is designed to non-selectively block alpha-adrenergic receptors and inhibit calcium entry into vascular smooth muscle cells thereby promoting relaxation of small arteries and arterioles and enhancing peripheral blood flow Buflomedil HCl exerts its biological activity primarily through non-selective alpha-adrenergic receptor blockade and inhibition of calcium influx Based on these pharmacological properties Buflomedil HCl holds research potential in the investigation of peripheral arterial disease ischemia intermittent claudication and vascular pathologies in experimental studies
Catalog No. 50-000-92231 Supplier Apexbio Technology LLC Supplier No. B18895.1
Buflomedil HCl is a peripheral vasodilating agent targeting alpha-adrenergic receptors It is designed to non-selectively block alpha-adrenergic receptors and inhibit calcium entry into vascular smooth muscle cells thereby promoting relaxation of small arteries and arterioles and enhancing peripheral blood flow Buflomedil HCl exerts its biological activity primarily through non-selective alpha-adrenergic receptor blockade and inhibition of calcium influx Based on these pharmacological properties Buflomedil HCl holds research potential in the investigation of peripheral arterial disease ischemia intermittent claudication and vascular pathologies in experimental studies