Learn More
Medchemexpress LLC Buflomedil hydrochloride | 35543-24-9 | 99.8% | 1 G
SDP
Catalog No. 5000406721
Encompass_Preferred
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. It can be used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Ki of 4.06 μM for α1A-AR
  • Ki of 6.84 μM for α1B-AR
  • Used for the study of peripheral circulatory disorders
Catalog No. 50-004-06721 Supplier Medchemexpress LLC Supplier No. HYB04841G
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. It can be used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Ki of 4.06 μM for α1A-AR
  • Ki of 6.84 μM for α1B-AR
  • Used for the study of peripheral circulatory disorders