Learn More
Medchemexpress LLC Buflomedil hydrochloride | 35543-24-9 | 99.8% | 1 ML
SDP
Catalog No. 5000409279
Encompass_Preferred
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. Its Kis are 4.06 μM for α1A-AR and 6.84 μM for α1B-AR. It can be used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Used for studying peripheral circulatory disorders
Catalog No. 50-004-09279 Supplier Medchemexpress LLC Supplier No. HYB048410MM/1ML
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. Its Kis are 4.06 μM for α1A-AR and 6.84 μM for α1B-AR. It can be used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Used for studying peripheral circulatory disorders