Learn More
Medchemexpress LLC Buflomedil hydrochloride | 35543-24-9 | 99.8% | 5 G
SDP
Catalog No. 5000409280
Encompass_Preferred
Buflomedil hydrochloride is a vasodilator agent that acts as an orally active α1A-adrenoceptor antagonist. It is utilized in the study of peripheral circulatory disorders and appears as a white to off-white solid.
  • Acts as an orally active α1A-adrenoceptor antagonist
  • Used for the study of peripheral circulatory disorders
  • Has a purity of 99.8%
  • Soluble in water and DMSO
Catalog No. 50-004-09280 Supplier Medchemexpress LLC Supplier No. HYB04845G
Buflomedil hydrochloride is a vasodilator agent that acts as an orally active α1A-adrenoceptor antagonist. It is utilized in the study of peripheral circulatory disorders and appears as a white to off-white solid.
  • Acts as an orally active α1A-adrenoceptor antagonist
  • Used for the study of peripheral circulatory disorders
  • Has a purity of 99.8%
  • Soluble in water and DMSO