Learn More
Medchemexpress LLC Buflomedil (hydrochloride) | 35543-24-9 | 99.8% | 500 MG
SDP
Catalog No. 5000409445
Encompass_Preferred
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. It is characterized by its Ki values of 4.06 μM for α1A-AR and 6.84 μM for α1B-AR. This compound is primarily used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Useful for studying peripheral circulatory disorders
  • High purity of 99.8%
  • Appears as a white to off-white solid
Catalog No. 50-004-09445 Supplier Medchemexpress LLC Supplier No. HYB0484500MG
Buflomedil hydrochloride is a vasodilator agent and an orally active α1A-adrenoceptor antagonist. It is characterized by its Ki values of 4.06 μM for α1A-AR and 6.84 μM for α1B-AR. This compound is primarily used for the study of peripheral circulatory disorders.
  • Vasodilator agent
  • Orally active α1A-adrenoceptor antagonist
  • Useful for studying peripheral circulatory disorders
  • High purity of 99.8%
  • Appears as a white to off-white solid