Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Buformin hydrochloride | 1190-53-0 | 99.78% | 50 MG
SDP

Catalog No. 5000411652
Encompass_Preferred

Buformin hydrochloride | 1190-53-0 | 99.78% | 50 MG

Catalog No. 50-004-11652 Supplier Medchemexpress LLC Supplier No. HYB2099A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Buformin hydrochloride | 1190-53-0 | 99.78% | 50 MG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.