Learn More
Medchemexpress LLC Buformin hydrochloride | 1190-53-0 | 99.8% | 1 ML
SDP
Catalog No. 5000409887
Encompass_Preferred
Buformin hydrochloride is a potent AMPK activator and an orally active biguanide antidiabetic agent. It decreases hepatic gluconeogenesis and lowers blood glucose production in vivo. It also exhibits anti-cancer activities and is used in cancer studies (e.g., cervical cancer and breast cancer).
  • Potent AMPK activator
  • Orally active biguanide antidiabetic agent
  • Decreases hepatic gluconeogenesis
  • Lowers blood glucose production in vivo
  • Has anti-cancer activities, applied in cancer studies (e.g., cervical cancer and breast cancer)
Catalog No. 50-004-09887 Supplier Medchemexpress LLC Supplier No. HYB2099A10MM/1ML
Buformin hydrochloride is a potent AMPK activator and an orally active biguanide antidiabetic agent. It decreases hepatic gluconeogenesis and lowers blood glucose production in vivo. It also exhibits anti-cancer activities and is used in cancer studies (e.g., cervical cancer and breast cancer).
  • Potent AMPK activator
  • Orally active biguanide antidiabetic agent
  • Decreases hepatic gluconeogenesis
  • Lowers blood glucose production in vivo
  • Has anti-cancer activities, applied in cancer studies (e.g., cervical cancer and breast cancer)