Learn More
Cayman Chemical Buformin (hydrochloride), Purity: ≥95%, CAS: 1190-53-0, 100 mg
SDP
Catalog No. 502968035
Encompass_Preferred
A biguanide that exhibits antihyperglycemic activity by increasing glucose utilization and inhibiting hepatic gluconeogenesis; inhibits the mitochondrial respiratory complex I and the mitochondrial ATP synthase; disrupts the unfolded protein response transcription program during glucose deprivation, inducing cell death in glucose-deprived tumor cells
Catalog No. 50-296-8035 Supplier Cayman Chemical Supplier No. 18507100 MG
A biguanide that exhibits antihyperglycemic activity by increasing glucose utilization and inhibiting hepatic gluconeogenesis; inhibits the mitochondrial respiratory complex I and the mitochondrial ATP synthase; disrupts the unfolded protein response transcription program during glucose deprivation, inducing cell death in glucose-deprived tumor cells