Learn More
Enzo Life Sciences Bufotalin (5mg). CAS: 471-95-4
SDP
Supplier:  Enzo Life Sciences ALX350282M005
Encompass_Preferred
Less potent Na+/K+-ATPase inhibitor than other bufadienolides. Potent immunosuppressor. Alternative name: (3beta,5beta,16beta)-16-(Acetyloxy)-3-,14-dihydroxybufa-20,22-dienolide, 3beta,14,16beta-Trihydroxy-5beta bufa-20,22-dienolide 16-acetate. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in 100% ethanol or chloroform. Long Term Storage: +4°C.
Catalog No. 50-200-8827