Learn More
Cayman Chemical BufuralolhydrochlorIde 10mg
SDP
Catalog No. 502967053
Encompass_Preferred
A non-selective antagonist of β-ARs that also has partial agonist activity; decreases mean arterial blood pressure and increases abdominal aortic blood flow in anesthetized cats at 0.3 and 1 mg/kg; has been used as a substrate to measure CYP2D6 activity
Catalog No. 50-296-7053 Supplier Cayman Chemical Supplier No. 1779410 MG
A non-selective antagonist of β-ARs that also has partial agonist activity; decreases mean arterial blood pressure and increases abdominal aortic blood flow in anesthetized cats at 0.3 and 1 mg/kg; has been used as a substrate to measure CYP2D6 activity