Learn More
Biolog Inc BUG AGAR 500GM
SDP
Catalog No. NC9152401
Encompass
Dehydrated Biolog Universal Growth Agar, 500g jar, makes approximately 435 plates
Catalog No. NC9152401 Supplier Biolog Inc Supplier No. 70101
Dehydrated Biolog Universal Growth Agar, 500g jar, makes approximately 435 plates