Learn More
FQS LLC Bug Alert Test Kit (AB/YM)
Catalog No. NC3663913
Encompass
Each kit contains 10 pairs of AB/YM tests.The Bug Alert Test comes with everything necessary to conduct 10 complete tests. This test can be used in diesel and gasoline. The Bug Alert Test is not endorsed by the Commercial Aviation Industry or other applications not listed.
Catalog No. NC3663913 Supplier FQS LLC Supplier No. FQS005
Each kit contains 10 pairs of AB/YM tests.The Bug Alert Test comes with everything necessary to conduct 10 complete tests. This test can be used in diesel and gasoline. The Bug Alert Test is not endorsed by the Commercial Aviation Industry or other applications not listed.