Learn More
Avalon International Corp Bug Check BF - 10 Bacteria/Mold Tests per Kit
Supplier:  Avalon International Corp BUGCHECKBF
Encompass
Bug Check BF; Simple easy to use test kit for counting total bacteria, fungi in all water based fluids; Examples: metalworking fluids, latex coatings, similar emulsions, adhesives, e coating paint systems, cooling tower water; 10/PK
Catalog No. NC9678807