Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ HT Protein Extraction Reagent

Catalog No. 709225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100mL
500mL
1L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-922-5 1L
70-922-3 100mL
70-922-4 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 70-922-5 Supplier MilliporeSigma™ Supplier No. 709225
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Eliminates common bioprocessing problems resulting from traditional cell lysis procedures

  • Combines BugBuster Protein Extraction Reagent and Benzonase™ Nuclease in one convenient reagent
  • Soluble proteins are gently extracted from E. coli without exposure to heat or oxidative damage
  • Viscosity is eliminated by nucleic acid digestion in a single step
  • The resulting protein extract can easily be fractionated by conventional purification techniques
  • Ideally suited for application in high throughput protein purifications
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1L
Content And Storage Store at +15° to +30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.