Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Bugbuster™ Master Mix
GSA_VA

Catalog No. 714564 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-456-4 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-456-4 Supplier MilliporeSigma™ Supplier No. 714564
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Conveniently combined extraction reagents in a single solution. Bugbuster™ Master Mix allows for maximum recovery of active soluble protein from both Gram-negative/Gram-positive bacteria.

  • BugBuster™ Extraction Reagent
  • Benzonase™
  • Nuclease
  • rLysozyme™
  • No need for dilution or separate addition steps.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 500 mL
Product Type Master Mix
Content And Storage 2 to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.