Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ BugBuster™ (primary amine-free) Extraction Reagent

Catalog No. 709233 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-923-3 100mL
70-923-4 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-923-3 Supplier MilliporeSigma™ Supplier No. 709233
Add to Cart
Add to Cart

Ideally suited for extraction of metal-dependent proteins

  • Special formulation of BugBuster designed for applications where primary amines would interfere if present in protein extract, such as protein immobilization or cross-linking
  • The PIPPS buffer used in BugBuster (primary amine-free) has a similar buffer capacity and pH range as the original Tris-buffered BugBuster, but will not complex metal ions, also making it ideally suited for extraction of metal dependent proteins
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100mL
Content And Storage Store at+15°C to+30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.