Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BugLoupe™ Magnifier
SDPSCI_ED

Catalog No. S65312 Shop All Carson Optical Products
Change view
Click to view available options
Magnification:
5x
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Magnification
S65312 5x
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65312 Supplier Carson Optical Supplier No. HU55
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

See the finest details of any insect.

  • Professional grade glass optics
  • Pre-focused for quick, easy viewing
  • Magnification: 4.5X

Specifications

Weight (Metric) 195.61 g
Product Type Magnifier, Bugloupe
Material Acrylic
For Use With (Application) To see the finest details of any insect
Weight (English) 6.9 oz.
Magnification 5x
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.