Learn More
Bachem Fmoc-Lys(palmitoyl-Glu-OtBu)-OH
SDP
Supplier:  Bachem 4080411.0001
Encompass_Preferred
Fmoc-Lys(palmitoyl-Glu-OtBu)-OH, 1491158-62-3, C46H69N3O8, Mr 792.07, 1g. Building block for GLP-1 analogs.
Catalog No. 50-194-7221