Learn More
Bachem Fmoc-Tyr(Me)-OH
SDP
Supplier:  Bachem 4042449.0005
Encompass_Preferred
Fmoc-Tyr(Me)-OH, 77128-72-4, C25H23NO5, Mr 417.46, 5g. Fmoc-4-methoxy-Phe-OH. Building block for oxytocin and vasopressin analogs as carbetocin, and for structure-activity studies.
Catalog No. 50-194-7353